Connect the back-pressure coil(s) between the CELL OUTLET port and the REGEN IN port. The back-pressure coils for 4 mm systems are provided in the Gas Separator Waste Tube Components Assembly (Item # 045825). The back-pressure coils for 2 mm systems are provided in the microbore tubing kit (Item # 052324).
CAUTION
The Dionex DRS 600, Dionex ERS 500e or Dionex AERS 500 Carbonate must be operated with the Gas Separator Waste Tube (Item # 045460).