Reagent-grade inorganic chemicals should always be used to prepare ionic eluents. Preferably, a lot analysis on each label will certify each chemical as meeting or surpassing the latest American Chemical Society standard for purity, a universally accepted standard for reagents.