Precise and accurate results require eluents free of ionic impurities; chemicals and deionized water used to prepare eluents must be pure, as described in this section. Low trace impurities and low particulate levels in eluents and regenerants also help protect the suppressor and system components from contamination. Suppressor performance is not guaranteed when the quality of the chemicals and water used to prepare eluents has been compromised.