See the following table for information on the instrument methods used with the DRS 600.
Mode | Description |
|---|---|
New method (ICS 6000 and Integrion) | When operating the DRS 600 under Legacy mode (Constant Current), you can create a new Instrument method from the CM Wizard and run the new method in a sequence. In the ICS 6000 Chromeleon Instrument Method Wizard, you can select the Legacy (CC) power mode. In the Integrion the mode is chosen or viewed as Constant Voltage or Constant Current, as shown in Operating the DRS 600 with Integrion with the Legacy Instrument Method. By matching the Mode as described in Operating DRS 600 suppressor with Chromeleon with the mode in the Instrument Method Wizard, the Legacy (Constant Current) operation can be pursued. |
Legacy instrument method (ICS 6000 and Integrion) | The DRS 600 operates in the legacy (constant current) mode by default. The legacy instrument method will operate without any error in the constant current mode for the entire sequence. The audit trail will capture the applied current information from the method. |