You can change the current mode for the ICS 6000 from Chromeleon. The current can be set from the System wizard or from the Instrument Method.
Procedure
  1. Press F8, or select the Settings icon.
  2. On the window that opens, right-click the Properties tab, then select View as Expert.
  3. View as expert
    View as expert
  4. On the left-side panel, select DC, then select Suppressor.
  5. Under Power Mode, select Legacy.
  6. Select legacy power mode
    Select legacy power mode