The default power mode is Legacy (constant current). Once the Legacy mode is selected, the DRS functions under constant current mode as shown in the following figure. The current can be set from the system wizard or from the Instrument Method.
To set optimum current setting for the suppressor, see Calculate the electrical current for the suppressor.
Procedure
- In the system ePanel, select the DC tab.
- In the Suppressor section, select Legacy from the Mode dropdown.