The power mode of operation for a Dionex DRS 600 suppressor is the Legacy Mode (Constant Current).
Procedure
  1. Establish the power mode of operation (Legacy/Constant Current is default).
  2. Create a new method from the Instrument Method Wizard and include this in a sequence.
  3. For most applications, using the wizard will be adequate to establish a DRS compatible method as described in Operate the DRS 600 with a method (Integrion and ICS 6000)