See the following table for the firmware and software requirements for the Dionex™ Integrion and Dionex™ ICS 6000 systems.
System | Minimum firmware | Chromeleon version |
|---|---|---|
Thermo Scientific™ Dionex™ Integrion™ HPIC™ System | Version 1.7.0 or later | Version 7.2.7 or later1) |
Thermo Scientific™ Dionex™ ICS 6000™ HPIC™ System | Included with system | Version 7.2.7 or later |