The Dionex DRS 600, Dionex ERS 500e and Dionex AERS 500 Carbonate suppressors are designed for different applications. See the following table for details.
Suppressor model | Suitable applications | Compatible ion chromatography system (ICS) |
|---|---|---|
Dionex DRS 600 | Direct replacement for:
Can be used in place of any of these suppressors where recycled eluent mode or external water mode is employed. | Any Dionex Ion Chromatography System (ICS) equipped with an analytical Anion or Cation exchange column set and an electrolytic suppressor controller, including:
NOTE: Some legacy systems require a standalone controller for installation. See Electrolytically regenerated suppressor control. |
Dionex ERS 500e | External water applications where difficult sample matrices are being analyzed. | |
Dionex AERS 500 Carbonate suppressor | Operation with carbonate and carbonate/bicarbonate eluents. Hardware design significantly improves noise performance with carbonate eluents without sacrificing performance or ease-of-use and delivers the lowest noise level of any electrolytic suppressor for carbonate eluents. | |
Dionex CRS 500 | Chemical suppression mode. |
CAUTION
The Dionex DRS 600, Dionex ERS 500e and Dionex AERS 500 Carbonate suppressors are not designed to be run on Dionex Capillary Ion Chromatography Systems (ICS-4000), or on Dionex Ion Chromatography Systems that do not have an electrolytic suppressor controller (Dionex ICS-90A, Dionex ICS-600, Dionex ICS-900).