The Dionex DRS 600, Dionex ERS 500e, and Dionex AERS 500 Carbonate should not be subjected to temperatures above 50°C for long durations during shipment, storage or operation, or for any duration above 80°C.
Ensure the suppressor is stored in a temperature-controlled environment away from direct exposure to sunlight or other sources of heat. Do not store the suppressor in environments where temperatures exceed 50°C, such as a parked car.