After neutralization, the sample can be fed into any Dionex Ion Chromatography system and operated in the concentrator mode. It is important that only low-pressure concentrator columns, such as the Dionex UTAC-XLP2 (Item # 072781) and Dionex TCC-XLP1 (Item # 063889), are used to protect the Dionex DRS 600 from damage.