The Dionex Dynamically Regenerated Suppressor (DRS) 600 operates in the constant current mode and uses the same clean ion exchange components (screens and membranes) as the previous model (Dionex ERS 500). The high-capacity of the Dionex DRS 600 offers high capacity suppression while adding minimal delay volume to the analytical system, which significantly expands the capabilities and simplifies the operation of ion chromatography.
The Dionex DRS 600 is available in two versions:
  • Anion Dynamically Regenerated Suppressor (Dionex ADRS™ 600): Provides continuous suppression of traditional eluents, and more concentrated eluents up to 200 mM NaOH.
  • Cation Dynamically Regenerated Suppressor (Dionex CDRS™ 600): Provides continuous suppression of concentrated eluents up to 100mN H2SO4 or MSA.
The Dionex DRS 600 system consists of the following:
  • Dynamically regenerated suppressor
  • Suppressor control
  • Back pressure coils
  • Gas separator waste tube
DRS and accessories
DRS and accessories
You can find information on all of the different support services of Thermo Fisher Scientific by navigating the following website:
https://www.thermofisher.com/us/en/home/technical-resources/contact-us.html