Sample | 100 µL of 22% phosphoric acid neutralized in the Dionex CDRS 600 |
Concentrator Column | Low Pressure Trace Cation Concentrator (i.e. Dionex IonPac TCC-XLP1, Item # 063889) |
Guard Column | Dionex IonPac CG12A Guard Column |
Analytical Column | Dionex IonPac CS12A Analytical Column |
Eluent | 20 mM Methanesulfonic acid (MSA) |
Eluent Flow Rate | 1.0 mL/min |
Electrolytic Suppressor | Dionex Cation Dynamically Regenerated Suppressor, Dionex CDRS 600 Recycled Eluent Mode |
or Chemical Suppressor | Dionex Cation Chemically Regenerated Suppressor, Dionex CCRS 500 |
Chemical Regenerant | 100 mN TBAOH |
Expected Background Conductivity | 20 mM MSA: <1 µS |
Expected System Operating Back Pressure | Without Guard: 1,400 psi (9.65 MPa) |
With Guard: 1,850 psi (12.75 MPa) |
Analyte | Conc. (µg/L) |
|---|---|
Sodium | 509.1 |
Ammonium | 438.6 |
Dimethylamine | B |
Potassium | 25.8 |
Trimethylamine | B |
Magnesium | 22.4 |
Calcium | 144.1 |