Sample | 100 µL of 20% NH4OH neutralized in the SP10 AutoNeutralizer |
Concentrator Column | Low-Pressure Trace Anion Concentrator (i.e., Dionex IonPac UTAC-XLP2, Item # 072781) |
Column | Dionex IonPac AG12A + Dionex IonPac AS12A |
Eluent | 2.7 mM Na2CO3/0.3 mM NaHCO3 |
Eluent Flow Rate | 1.5 mL/min |
Electrolytic Suppressor | Dionex Anion Dynamically Regenerated Suppressor, Dionex ADRS 600 Recycled Eluent Mode |
or Chemical Suppressor | Dionex Anion Chemically Regenerated Suppressor, Dionex ACRS 500 |
Chemical Regenerant | 50 mN H2SO4 |
Expected Background Conductivity | 14–16 µS |
Analyte
- 1. Chloride
- 2. Bromate
- 5. Nitrate
- 7. Sulfate
- 8. Oxalate