The following examples demonstrate sample neutralization can be performed with the Thermo Scientific Dionex IonPac AS12A column using the ADRS 600 suppressor
Sample | 100 µL of 25% NaOH neutralized in the SP10 AutoNeutralizer |
Concentrator Column | Low-Pressure Trace Anion Concentrator (i.e., Dionex IonPac UTAC-XLP2, Item # 072781) |
Column | Dionex IonPac AG12A + Dionex IonPac AS12A |
Eluent | 2.7 mM Na2CO3/0.3 mM NaHCO3 |
Eluent Flow Rate | 1.5 mL/min |
Electrolytic Suppressor | Dionex Anion Dynamically Regenerated Suppressor, Dionex ADRS 600 Recycled Eluent Mode |
or Chemical Suppressor | Dionex Anion Chemically Regenerated Suppressor, Dionex ACRS 500 |
Chemical Regenerant | 50 mN H2SO4 |
Expected Background Conductivity | 14–16 µS |
Peak Assignment/Sample Concentration | See the table below. |
| Spiking Standard Conc. | 25% NaOH Conc. | Expected Spiked Value Conc. | 25% NaOH Spiked Conc. |
|---|---|---|---|---|
Chloride | 0.100 | 0.155 ± 0.003 | 0.25 | 0.258 ± 0.004 |
Bromide | 0.100 | ND2) | 0.100 | 0.099 ± 0.008 |
Chlorate | 0.100 | ND | 0.100 | 0.099 ± 0.009 |
Nitrate | 0.100 | 0.011 ± 0.006 | 0.111 | 0.199 ± 0.013 |
Phosphate | 0.100 | 0.106 ± 0.025 | 0.206 | 0.183 ± 0.017 |
Sulfate | 0.200 | 0.348 ± 0.004 | 0.548 | 0.553 ± 0.011 |
Oxalate | 0.100 | 0.134 ± 0.028 | 0.234 | 0.238 ± 0.022 |