Sample | 100 µL of 10% NaOH neutralized in the SP10 AutoNeutralizer |
Concentrator Column | Low-Pressure Trace Anion Concentrator (i.e., Dionex IonPac UTAC-XLP2, Item # 072781) |
Trap Column | Anion Trap Column (i.e., Dionex IonPac ATC-HC 500, Item # 075978) |
Guard Column | Dionex IonPac AG11 Guard Column |
Analytical Column | Dionex IonPac AS11 Analytical Column |
Eluents | E1: Type I Deionized Water |
E2: 5.0 mM NaOH | |
E3: 100 mM NaOH | |
Eluent Flow Rate | 2.0 mL/min (4-mm) |
Electrolytic Suppressor | Dionex Anion Dynamically Regenerated Suppressor, Dionex ADRS 600 Recycled Eluent Mode |
or Chemical Suppressor | Dionex Anion Chemically Regenerated Suppressor, Dionex ACRS 500 |
Chemical Regenerant | 50 mN H2SO4 |
Expected Background Conductivity | 0.5 mM NaOH: 1 µS |
35 mM NaOH: 3.5 µS | |
Expected System Operating Back Pressure | Without Guard: 1,600 psi (11.03 MPa) |
With Guard: 1,850 psi (12.75 MPa) |
| Standard Conc. | 10% NaOH Conc. | Expected Spiked Value Conc. | 10% NaOH Spiked Conc. |
|---|---|---|---|---|
Quinate | 1.000 | ND1) | 1.000 | 1.435 |
Fluoride | 0.200 | 0.030 | 0.230 | 0.2444 |
Acetate | 1.000 | 0.081 | 1.081 | 1.301 |
Propionate | 1.000 | ND | 1.000 | 1.069 |
Formate | 1.000 | 0.213 | 1.213 | 1.216 |
Methylsulfonate | 1.00 | ND | 1.000 | 1.070 |
Pyruvate | 1.000 | ND | 1.000 | 1.103 |
Valerate | 1.000 | ND | 1.000 | 1.009 |
Monochloroacetate | 1.000 | ND | 1.000 | 0.811 |
Bromate | 1.000 | ND | 1.000 | 1.036 |
Chloride | 0.400 | 8.750 | 9.150 | 9.095 |
Trifluoroacetate | 1.000 | ND | 1.000 | 1.055 |
Bromide | 0.600 | ND | 0.600 | 0.640 |
Nitrate | 0.600 | 0.048 | 0.648 | 0.657 |
Chlorite | 0.600 | 1.178 | 1.778 | 1.870 |
Selenite | 1.000 | ND | 1.000 | 0.990 |
Carbonate | --- | --- | --- | --- |
Malonate | 1.000 | ND | 1.000 | 1.303 |
Maleate | 1.000 | ND | 1.000 | 1.026 |
Sulfate | 1.000 | 9.842 | 10.842 | 10.925 |
Oxalate | 1.000 | 0.166 | 1.166 | 1.244 |
Tungstate | 2.000 | ND | 2.000 | 2.201 |
Phthalate | 2.000 | ND | 2.000 | 2.086 |
Phosphate | 2.000 | ND | 2.000 | 2.120 |
Chromate | 2.000 | ND | 2.000 | 2.287 |
Citrate | 2.000 | ND | 2.000 | 2.214 |
Tricarballylate | 2.000 | ND | 2.000 | 2.098 |
Isocitrate | 2.000 | ND | 2.000 | 2.112 |
cis-Aconitate | 2.000 | ND | 2.000 | 1.512 |
trans-Aconitate | 2.000 | 0.072 | 2.072 | 2.303 |