If a sample contains transition metals in ppm (mg/L) levels, these elements may interfere with cation detection by suppressed conductivity. The addition of a complexing agent such as pyridine-2,6-dicarboxylic acid (PDCA, Item # 039671) to the CS12A eluent is required to selectively remove transition metals from the eluent via the suppression system.
Anion | ppb1) | Cation | ppb2) |
|---|---|---|---|
Chloride | 4 | Lithium | 0.03 |
Bromide | 20 | Sodium | 1.0 |
Chlorate | 30 | Potassium | 2.0 |
Nitrate | 20 | Magnesium | 0.6 |
Phosphate | 50 | Calcium | 0.8 |
Sulfate | 30 | ||
Oxalate | 50 |