The following examples are presented to highlight the analysis of trace anions or cations found in the concentrated matrices that have been treated using a Dionex Dynamically Regenerated Suppressor (Dionex DRS 600) in Neutralizer Mode.
The analysis may be done on any one of several Dionex anion or cation exchange column sets. Please refer to the Product Manual for the anion or cation exchange column set installed in your system for detailed operation and troubleshooting information.
Because of its low operating pressure, the Dionex IonPac UTAC-XLP2 (Item # 072781) or Dionex IonPac TCC-XLP1 (Item # 063889) should be used to concentrate the trace anions or cations in the sample neutralized by the Dionex DRS 600. The use of other concentrators may result in excessive back pressure to the Dionex DRS 600, thus damaging the suppressor.
NOTE
The Dionex AERS 500 Carbonate suppressors are not recommended for use with the Neutralization Mode of Operation. The Dionex DRS 600 or ERS 500e should be used.