You can open a result file from multiple locations:
Procedure
- Do any one of the following to open the result file generated by the analysis:
- From the Job Queue page: If the Job Queue page is open, double-click the row for the completed analysis.
- From the Analysis Results page of a study: If the Study page is open, select the Analysis Results tab. Then, double-click the row for the completed analysis.
- From the Start page: If you closed the study after the run completed, select the result file name under Recent Results on the Start Page.
- From the menu bar: If the Start page is closed, open it by selecting View > Start Page from the menu bar.
- The result file opens as a tabbed document in the application window.