In this tutorial, you will add the eleven raw data files in the CD 3.5 Example Studies\LC\Stable Isotope Labeling folder to the study.
Refer to the previous Locate the example files topic for more information on locating the raw data files used in this tutorial.
Procedure
  1. On the Input File Selection page of the wizard, select Add Files.
  2. Browse to the file location where you copied the example data.
  3. Select the eleven raw data files in the Stable Isotope Labeling folder and select Open.
  4. In the wizard, the file names of the selected files appear and the Next button becomes available.
  5. RAW files (11) selected
    Figure RAW files (11) selected
  6. Select Next to continue.
  7. The Input File Characterization page of the wizard opens.
  8. Imported files with assigned file numbers
    Figure Imported files with assigned file numbers