For the stable isotope labeling analysis, the Compounds table contains up to 37 visible table columns. With the Stable Isotope Labeling layout applied, the Compounds table contains 20 visible table columns (see Apply the Stable Isotope Labeling layout).
To select the table columns that you want to export to a spreadsheet file, do the following:
Procedure
- Select the Field Chooser
icon for the Compounds table.
- In the Field Chooser dialog box, clear the checkboxes for the columns that you do not want to export to a spreadsheet file. For this tutorial, clear all the checkboxes except the following five columns:
Name, Area (Max), Calc. MW, Formula, and RT - Close the Field Chooser dialog box.