To filter the Compounds table by the best mzCloud match, do the following:
Prerequisites
  1. The Compounds table in the example result file is filtered by the exchange rate as described in Filter the Compounds table by the relative exchange rate.
Procedure
  1. Open the Result Filters view if it is not open.
  2. Select Add Property below the last Rel Exchange [%] filter.
  3. In the dropdown property list, select mzCloud Best Match.
  4. In the pink dropdown conditions list to the right, select Is Greater Than or Equal To.
  5. In the value box, enter 99.
  6. Select Apply Filters.
  7. The Compounds table is now reduced to ten compounds.
  8. Close the Result Filters view by selecting the Close icon in the upper‑right corner.
  9. See Select columns that you want to export.