To filter the Compounds table by the relative exchange rate of each compound, do the following:
Procedure
  1. Select the Compounds tab for the main Compounds table to make it the active table.
  2. From the application menu bar, select View > Result Filters
  3. The Result Filters view opens as a floating window. Because the processing workflow included the Mark Background Compounds node and the Analyze Labeled Compounds node, the filter for the Compounds table already includes a filter for background compounds and a filter for compounds without a formula. The figure below shows the default filters for the example result file.
  4. On the right side of the Result Filters view, set up filters for the relative exchange rate by following these steps for each of the three labeled input files:
  5. Select Add Property, and then select Rel. Exchange [%] from the list.
  6. In the pink relation list, select Is Greater Than or Equal To.
  7. In the value box next to the relation list, enter 98.
  8. In the pink condition list, select In File.
  9. In the Green sample list, select one of the labeled input files.
  10. Repeat step 3a through step 3e to add filter all the labeled input files.
    The figure shows the filter set.
  11. Select Apply Filters.
  12. The applied filter set reduces the number of visible rows in the Compounds table to 248.