To filter the Compounds table by the selected items, do the following:
Procedure
  1. If the Compounds table is not the active table, select its tab to make it active.
  2. Manually select the check boxes for the compounds of interest.
  3. From the menu bar, select View > Result Filters.
  4. Because the processing workflow included the Mark Background Compounds node and the Analyze Labeled Compounds node, the Compounds table is currently filtered by two properties—Background and Formula.
  5. Select Add Property and select Checked.
  6. Select Apply Filters.
  7. The Compounds table displays only the selected compounds.
  8. To undo the Checked filter, select Remove to its right. Then, select Apply Filters again.
  9. Now that you have removed the Checked filter, the Compounds table contains the original set of compounds.
  10. See Filter the Compounds table by the relative exchange rate.