In this section of the tutorial, you will investigate a contaminating mass in an unlabeled sample:
Prerequisites
- You are viewing the Stable Isotope Labeling
- The Compounds table is sorted by Area (Max.) in descending order.
Procedure
- In the Compounds table, select the pin icon to left of row #84 to pin the compound (with a Calc. MW of 248.17486 and RT of 11.048 min) to the top of the table.
- The row for the compound is pinned to the top of the table, allowing you to scroll down while keeping the row visible.
- In the Labeling Status column, select the expand icon for the column.
Figure Compound with Calc. MW of 248.17486 (row #84) in the Compounds table (with the Name and Labeling Status columns frozen at the left). - The row headers appear. Because you grouped the samples by sample type, the samples are also grouped by sample type in the Labeling Status column.
- The unlabeled samples (F2, F3, and F4) are denoted with a red box, indicating the presence of a contaminating mass in these samples.
- Select row #84 (if necessary) and then below the Compounds table, select Show Related Tables.
- The first-level related tables appear: Structure Proposals, Compounds per File, Predicted Compositions, Features, Labeled Compounds per File, Formulas, Metabolika Results, mzCloud Results, ChemSpider Results, and Metabolika Pathways.
- With the Stable Isotope Labeling layout applied, the Labeled Compounds per File table is selected by default.
- In the Labeled Compounds per File table, do the following:
- Right-click the table and select Enable Column Fixing. Then, select the pin icon for the Exchange Rate [%] column.
- Select the expand icon for the Exchange Rate [%] column.
- The row headers appear.
- The Exchange Rate [%] column shows that the contaminating mass is possibly a compound with a mass of M+9.
Figure Labeled Compounds per File table for the compound with Calc. MW of 248.17486 - Refer to View a trendline chart or box-and-whiskers plot to continue the tutorial.