In this section of the tutorial, you will review the exchange rates for L-glutamic acid, a compound detected across all sample files in the analysis.
TIP
In this tutorial, the row numbers listed for the example compounds are accurate only when the Compounds table is sorted by Area [Max.] in descending order, where row #1 describes gamma-glutamylcysteinylglycine and row #2 describes L-glutamic acid.
To sort the compounds by Area [Max.] in descending order, select the Area [Max.] column header until the sort arrow is pointed down.
Prerequisites
- You must apply the Stable Isotope Labeling layout to the result file.
Procedure
- In the Compounds table, select the pin icon to the left of L-glutamic acid (row #2).
- The row for L-glutamic acid is pinned to the top of the table, allowing you to scroll down while keeping the L-glutamic acid row visible.
Figure L-glutamic acid pinned at the top of the Compounds table - Then, pin the Name column at the left of the Compounds table as follows:
- Right-click the Compounds table and select Enable Column Fixing.
Figure Shortcut menu for the Compounds table - Pin icons appear to the right of the column headings.
- Select the pin icon to the right of the Name column heading (pinned (
) and unpinned (
)).
- The Name column is pinned to the left of the table, allowing you to scroll to the right while keeping the Name column visible.
- Scroll to the right of the Compounds table to view the Rel. Exchange [%] column.
- To view the input file names in the Rel. Exchange [%] column, do either of the following:
- Select the expand
icon next to the column header.
- Right‑click the Compounds table and select Expand All Column Headers to expand all the table column headers.
- The relative exchange rate for L-glutamic acid is 98% for the labeled samples (F9, F10, and F11) and 0% for the unlabeled samples (F2, F3, and F4).
Figure Relative exchange rates for L-glutamic acid - To view the exchange rate for the isotopologues of L-glutamic acid, do the following:
- In the Compounds table, select the row for L-glutamic acid (row #2, pinned at the top of the table).
- Below the Compounds table, select Show Related Tables.
- The first-level related tables appear: Structure Proposals, Compounds per File, Predicted Compositions, Features, Labeled Compounds per File, Formulas, Metabolika Results, mzCloud Results, ChemSpider Results, and Metabolika Pathways.
- With the Stable Isotope Labeling layout applied, the Labeled Compounds per File table is selected by default.
- In the Labeled Compounds per File table, scroll to the Exchange Rate [%] column.
- To view the isotopologues, select the expand
icon next to the Exchange Rate [%] column heading.
Figure Exchange Rate and Study File columns in the Labeled Compounds per File table for L-glutamic acid - The Exchange Rate [%] column contains 25 sub-columns because the analysis specified a maximum exchange of 25 atoms for any of the detected compounds in the Analyze Labeled Compounds node > Label Settings area: Max Exchange = 25 (default).
- The exchange rates for the labeled samples are 92% for the 13C5H9NO4 isotopologue and 7% for the 13C4CH9NO4 isotopologue of L-glutamic acid.
- Irrelevant sub-columns for unprocessed elemental compositions have a gray background.
- Sub-columns for isotopologues have a pink to red background that darkens as the exchange rate increases.
- To view the exchange rates for the adducts of L-glutamic acid, do the following:
- In the Labeled Compounds per File table, select the row for F11, one of the labeled samples.
- Below the Labeled Compounds per File table, select Show Related Tables.
- The second-level of related tables appear: Compounds, Labeled Features per File, and Input Files.
- In the second-level Related Tables area, select the Labeled Features per File tab.
- The Exchange Rate [%] column shows the exchange rates for the pseudo molecular ion [M+H]+1.
Figure Labeled features for L-glutamic acid in study file F11 - Refer to Review the labeling status to continue the tutorial.