In addition to the standard default result file layout, the application includes two additional result layouts: Statistics and Stable Isotope Labeling.
For this tutorial, apply the Stable Isotope Labeling layout as follows:
Procedure
  1. From the application menu bar, select Window > Apply Layout > Stable Isotope Labeling.
  2. Applying the Stable Isotope Labeling layout does the following to the example result file:
Table Changes in the result tables after applying Stable Isotope Labeling layout
Action
Details
Hides the following main tables.
  • Compounds per File
  • Features per File
  • ChemSpider Results
  • Labeled Compounds per File
  • Labeled Features per File
  • Metabolika Results
Hides the following columns in the Compounds table
  • #Adducts
  • #Metabolika Pathways
  • #Scan IDs (mzCloud)
  • Annot. Δ Mass [Da]
  • Annotation MW
  • Avg. Exchange
  • Background
  • FISh Coverage
  • ID
  • Metabolika Pathways
  • MS Depth
  • mzCloud Library Matches
  • PQF: Area Ratio
  • PQF: FWHM2Base
  • PQF: Gaps Ratio
  • PQF: Jaggedness
  • PQF: Modality
  • PQF: Number of Gaps
  • PQF: Number of Points
  • PQF: Zig-Zag Index
  • Structure
Displays the following columns in the Compounds table
  • #ChemSpider Results
  • #mzCloud Results
  • Annot. Source
  • Annot. D Mass [ppm]
  • Area
  • Area (Max).
  • Calc. MW
  • Checked
  • Comments
  • Formula
  • Labeling Status
  • MS2
  • MS2 Purity [%]
  • mzCloud Best Match
  • mzCloud Best Match Confidence
  • Name
  • Peak Rating
  • Peak Rating (Max.)
  • Rel. Exchange [%]
  • Polarity
  • RT [min]
  • Tags
Opens the Related Tables pane to the Labeled Compounds per File table for the first compound in the Compounds table.
Opens the Isotopologues Distribution Chart, Trend Chart, and Metabolika Pathways views as tabbed views on the bottom right of the page.
Selects the Rel. Exchange data property for the Trend Chart view.
Figure Results page after applying the Stable Isotope Labeling layout
Results page after applying the Stable Isotope Labeling layout