To open the reprocessed analysis result, do the following:
Procedure
- In the Job Queue, double-click the row for the completed Pesticides with ID.cdResult analysis.
- The result file opens as a tabbed document in the application window.
TIP
If you did not reprocess the original result file, you can use the Pesticides with ID.cdResult file provided on the Compound Discoverer 3.5 media.
TIP
To make sure that the result page is set to the default layout, select Window > Reset Layout from the application menu bar.