In this section of the tutorial, you will select the analysis results file for reprocessing.
TIP
You can use either of the following study files to continue this tutorial:
  • The Pesticides.cdStudy file that you generated by creating your own study and running a core GC CI analysis
  • The example Pesticides.cdStudy file provided on the Compound Discoverer 3.5 media: Example Studies\GC\PCI\Pesticides\Pesticides.cdStudy
Procedure
  1. Open the Pesticides study file by doing one of the following:
  2. From the application Start page, select the link for the Pesticides.cdStudy file under Recent Studies.
  3. From the application menu bar, select File > Open Study to locate and open the Pesticides.cdStudy file.
  4. On the study page, select the Analysis Results tab.
  5. The Analysis Results page opens, showing one analysis result file for the study: Pesticides Core.cdResult.
  6. To reprocess the result file, do one of the following:
  7. Right-click the result and select Reprocess from the shortcut menu.
  8. Select the result and select Reprocess on the command bar above the Analysis Results table.
  9. The Analysis pane reopens and the Processing Step status bar changes to Partially Reprocessing.
  10. The Workflows page populates with the current processing workflow.
  11. In the Analysis pane, the Files for Analysis area populates with the input files for the analysis.
  12. In the Analysis pane, the Run button remains unavailable until you edit the analysis.
  13. Once you edit any of the parameter settings in the core workflow nodes or add or remove input files, the Processing Step status changes to Fully Reprocessing.
  14. Refer to Add downstream nodes to the processing workflow to continue the tutorial.