To check if the instrument is ready, follow these steps:
Procedure
  1. In the Chromeleon Console, under the Instruments category, select the instrument by clicking its name in the navigation pane.
  2. In the work area, select the ePanel tab for the required module. For example, to check the baseline, select the detector’s ePanel and select the Monitor Baseline button.