A report template is a spreadsheet-like file that defines how data is displayed, printed, or exported.
To create a report template, follow these steps:
Procedure
- In the Chromeleon Console, from the Create menu, select Report Template.
- Select one of the predefined templates and then select OK.
- Select a file location and enter a file name. Select OK to save the report template and close the wizard.
- The report template opens in the Chromatography Studio under the Report Designer category.
TIP
For more information about creating a report template, refer to the Designing Reports: Open a Report or Create a Report Template topic in the Chromeleon 7 Help.