A processing method is a set of parameters used to analyze raw chromatography data. A processing method typically includes signal processing settings, peak detection parameters, a defined set of components with peak identification parameters, and calibration settings.
To create a processing method, follow these steps:
Procedure
  1. In the Chromeleon Console, from the Create menu, select Processing Method.
  2. Enter a name, select a file location, and enter a comment (adding a comment is optional).
  3. Click Finish to save the processing method and close the wizard.
  4. The new processing method opens in the Chromatography Studio.
TIP
For more information about creating a processing method, refer to the Creating/Modifying Processing Methods: Create a Processing Method topic in the Chromeleon 7 Help.