Before you can control an instrument, you must connect the modules to the Chromeleon Instrument Controller.
To connect the modules, follow these steps:
Procedure
- In the Chromeleon Console, select the Instruments category and then select your instrument’s name in the navigation pane.
- In the work area, select the ePanel tab for the required module (for example, Pump).
- Depending on the user interface of the instrument, do one of the following:
- Select the Connect check-box.
- Drag the slider bar to connect the module or instrument to Chromeleon.
- The green color indicates that a connection has been established.
TIP
For more information about connecting your instruments, refer to the Controlling Instruments in the Console topic in the Chromeleon 7 Help.