If the Instrument Controller service is stopped, start it by following these steps:
Procedure
  1. Right-click the Chromeleon tray icon in the notification area of the Windows taskbar.
  2. Select Start Chromeleon Instrument Controller.
For more information about the Chromeleon Services Manager see Chromeleon Services Manager.