After data has been acquired and processed, you can review and report results.
Figure Creating a report in Chromeleon
Sequence contains an associated report templateUse existing Thermo Scientific templatesCreate a new report template in the Report DesignerSave, export, or print a reportnoyesnoyesCreate a custom report template by adding spreadsheets, tables, graphs, and variablesSelect an existing report templateModify an existing report template
You can create report templates (which output results and other data) in any format and layout that you require, or use one of Chromeleon’s default templates (see Reviewing and reporting results). To review, print, and export results, you must first associate a template with the sequence.
In Chromeleon, you can create an electronic report with worksheets containing chromatograms, tables, charts, calibration curves, and other values. You can require electronic reports to go through an electronic signature process for approval.