The Chromeleon Administration Console is the central access point for all administrative tasks in Chromeleon. In the Chromeleon Administration Console, you can do the following:
  • Manage users
  • Allocate licenses
  • Plan, and organize tasks with the Scheduler
  • Define global policies and organizational units
  • Manage data vaults
  • Configure the Discovery Service
  • Monitor other Chromeleon services
  • Manage domain resources
The Administration Console window is divided into four areas (see Figure 2: Administration Console).
Figure Administration Console window
1234
No.DescriptionNo.Description
1
Title bar: You can right-click anywhere in the title bar to minimize, maximize, restore, move, or re-size the Administration Console window.
3
Status bar: On the left, it displays the name of the current user. On the right, it displays the status of the Chromeleon domain controller and the username of the current user.
2
Menu bar: Use this bar to browse and select items.
4
Work area: Displays content details for the item selected in the menu bar.
TIP
For more information about the Administration Console, refer to the Administering Chromeleon topic in the Chromeleon 7 Help.