When printing results, you can do either of the following:
Procedure
TIP
With the autorepeat area function, you can create a single report template that is suitable for a range of methods (regardless of whether a method uses one data channel or several, or whether it analyzes only one compound or several). Chromeleon automatically reports the relevant results. For details, refer to the Define Autorepeat Areas topic in the Chromeleon 7 Online Help.
To print the results for the currently selected injection, follow these steps:
Procedure
- In the Chromatography Studio, select the Report Designer category.
- In the navigation pane, select the injection that you want to print.
- From the Chromatography Studio menu, select Print.
- The Print dialog box opens.
- In the Print dialog box, do the following:
- Select the Apply to Current Injection option.
- Select the printer that you want to use.
- Select the sheets that you want printed.
- Select Print.
- The results for the current injection are now being printed.
To print results for an entire sequence, follow these steps:
Procedure
- In the Chromatography Studio, select the Report Designer category.
- In the navigation pane, select the injection that you want to print.
- From the Chromatography Studio menu, select Print.
- The Print dialog box opens.
- In the Print dialog box, do the following:
- Select the Apply to Current Sequence option.
- Select the printer that you want to use.
- Select the sheets that you want printed. Select the down arrow
in the Condition column to apply print conditions for the selected sheet.
- Select Print.
- The results for the current sequence are now being printed.
TIP
To print several injections from a sequence, first select the injection row or rows that you want to print, right-click anywhere among the selected rows, and then select Print.
TIP
For more information about printing the results, refer to the Printing Reports: Print the Report for a Sequence topic in the Chromeleon 7 Help.