During analysis, it is useful to monitor the detector signal and other instrument parameters (such as pressure and temperature).
To monitor an ongoing analysis, do the following:
Procedure
- In the Chromeleon Console, under the Instruments category, select your instrument’s name in the navigation pane.
- Select the Home ePanel in the work area to monitor the detector signal and audit trail.
NOTE
The settings of other modules can be monitored on their respective ePanels.