To check if the instrument is ready, follow these steps:
Procedure
- In the Chromeleon Console, under the Instruments category, choose the instrument by selecting its name in the navigation pane.
- In the work area, select the ePanel tab for the required module. For example, to check the baseline, select the detector’s ePanel and select the Monitor Baseline button.