A processing method is a set of parameters used to analyze raw chromatography data. A processing method typically includes signal processing settings, peak detection parameters, a defined set of components with peak identification parameters, and calibration settings.
To create a processing method, follow these steps:
Procedure
- In the Chromeleon Console, from the Create menu, select Processing Method.
- Enter a name, select a file location, and enter a comment (adding a comment is optional).
- Select Finish to save the processing method and close the wizard.
- The new processing method opens in the Chromatography Studio.
TIP
For more information about creating a processing method, refer to the Creating/Modifying Processing Methods: Create a Processing Method topic in the Chromeleon 7 Help.