The Chromeleon 7 Help provides useful information to help you work with the Chromeleon 7 chromatography software (see Chromeleon 7 Help). You can open the Chromeleon 7 Help either from the Help menu or from any dialog box or area that you are currently working in (referred to as "context-sensitive help"). If you open the Help from the Help menu, you can browse the information by using the Contents, Index, or Search tabs. If you open the Help from the dialog box or area that you are working in, the topic of interest opens directly.
You can open context-sensitive Help in one of the following ways:
Procedure
- Press the F1 key while you are in the portion of the software that you want to access the Help for.
- Select the question mark (
) in the upper-right corner of a dialog box that you want to access the Help for.
- Select the Help button in an active wizard page that you want to access the Help for.
NOTE
Not all wizard pages have a Help button.