Chromeleon services run in the background without a direct user interface. The status of each service is indicated by the icon displayed next to the name. The icons are displayed as follows:
  • The green icon indicates a running service.
  • The grey icon indicates a stopped service.
  • The orange icon indicates that the service is starting or stopping.
  • The black icon indicates that the service is not installed or cannot be reported.