The Chromeleon Instrument Controller service handles all real-time instrument processes (communications, control, and data acquisition). It runs in the background as a Windows service on PCs that are physically connected to instruments. The status of this service is represented by an icon ( or
) in the notification area of the Windows taskbar.