The Post Installation Configuration is automatically executed at the end of the Chromeleon setup. Thus, it is separated from the core installation of Chromeleon. In case of errors in the Chromeleon configuration, fix the problem and re-run the post installation program.
CAUTION
Setup settings as defined in the ResponseFile.xml are not taken into consideration when you re-run the post installation program.
- If you wish to enable or disable certain installation tasks when re-running the post installation program, you must use command-line options.
The Post Installation Configuration includes several configuration tasks, such as:
- Configuring the license
- Connecting to a Chromeleon domain
- Installing standard local data vaults
- Upgrading local data vaults to the current database schema
You can also execute it as a standalone application (PostInstallation.exe) after the Chromeleon installation:
Procedure
- Navigate to C:\Program Files (x86)\Thermo\Chromeleon\bin.
- Click PostInstallation.exe to execute the Post Installation Configuration application.
If you want to enable or disable tasks run by the Post Installation Configuration, you must use command-line options.