The Chromeleon setup program (Install.exe) accepts command-line options and properties. These can be used to trigger an unattended installation and to fine-tune the installation and the configuration process.
Properties
Properties are defined via the command line using the following syntax:
PROPERTY=PropertyValue
The following table shows the available properties.
Property
Description
InstallFolder
Specifies the installation folder.
The default installation folder (64-bit computers) is C:\Program Files (x86)\Thermo\Chromeleon\.
LogFilesBackupDir
Specifies the backup location of the setup log files.
The default location is C:\ProgramData\Dionex\Chromeleon\SetupLogs.
Note: Using a different location will prevent the System Status Report tool from including the setup log files for troubleshooting.
WaitTimeMinutesForSequenceQueuesIdle
The Chromeleon Setup will abort the installation if an active instrument queue is detected. Use this property to specify a wait time (minutes) for the instrument queue(s) to become idle.
Chromeleon will check for active queues every minute until the wait time has elapsed:
  • If there are still active queues, the installation will be aborted.
  • If no active queues can be detected, the installation will be continued.
NOTE
Property names are case sensitive.
Switches
In addition to properties, the setup program (Install.exe) also accepts numerous “switches”.
The following table shows the available command-line switches.
Property
Description
/?, /HELP
Lists all switches supported by Chromeleon.
<no option>,
/package,
/update
Installs or configures the product using the default settings.
/repair
Repairs the product (or installs if not installed).
/uninstall
Uninstalls the product.
/q, /quiet,
/s, /silent
Installs or configures the product in quiet mode without user interaction.
/promptrestart
Prompts the user for restart if necessary (default, recommended).
When used with the /q or /s switch, the prompt will be suppressed, and the computer will be restarted automatically when needed.
/forcerestart
Always restarts the computer after installation.
/norestart
Does not restart after the installation is complete. This option is not recommended.
/ib,
/ignoreblockers
Ignores all blockers and forces setup to continue. Not recommended.
Blockers are circumstances that the setup considers critical and would therefore stop.
/l <path\file>, /log <path\file>
Logs the installation events to the specified file/location (default location is the %TEMP% folder.)
/c:<argument>, /config:<argument>
Determines the system configuration tasks.
/sc, /skipconfig
Skips the system configuration phase.
/sq, /skipiq
Skips the installation qualification phase.
/lc, /listcomponents
Lists all selectable components. The installation of these components can be enforced or suppressed using a response file.
/rf:<path\file>, /responsefile:<path\file>
Specifies the response file to be used.