NOTE
The restrictions described below are only applicable for IPCs. They are also only relevant for customers who use Thermo Fisher USB drivers for IC, LC, A2D, and TRACE1110 equipment.
Do not use unattended (silent) mode if you are installing Chromeleon 7 for the first time on a new Windows computer. Doing so will prevent the Chromeleon USB driver package from being installed.
To install the Chromeleon USB driver package:
Procedure
- Select Always trust software from “Thermo Fisher Scientific” to confirm the Windows Security prompt.
- Click Install.
This prompt is only issued during routine installation of Chromeleon 7 using the Chromeleon setup kit.
NOTE
The Chromeleon USB Driver package can be installed in unattended mode on an IPC in the following cases:
- You are upgrading an existing installation of Chromeleon where the Windows security prompt for the Chromeleon USB Driver package was previously confirmed as described above.
- You are using image deployment to install Chromeleon, and installation of the Chromeleon USB Driver package was confirmed on the reference computer before the image file was created.