NOTE
You need access to the Chromeleon installation medium to repair an installation.
Procedure
- Insert the Chromeleon installation medium into the PC.
- Click Run Install.exe.
- This starts the Chromeleon setup in Maintenance mode.
- Click Repair to run a repair installation.
- Follow the on-screen instructions to repair the installation.