Thermo Fisher periodically releases Maintenance Updates (MU) between Service Releases to provide a greater level of stability for customer environments. Maintenance Updates typically only include bug fixes but do not provide any new functionality. Install Chromeleon maintenance updates to the latest version to ensure stability of the software.
To install a Chromeleon maintenance update, an installed Chromeleon main or service release is required. The maintenance update can only be installed over the respective main or service release it is based on.
If not yet present, install the Chromeleon main or service release as described in this document.
To install the maintenance update:
Procedure
- Insert the maintenance update installation medium into your PC.
- The AutoPlay window displays.
- Click Run Install.exe to start the installation procedure.
- Read and accept the Chromeleon license terms and conditions.
- Click Install to start the installation. The setup program is executed. Follow the on-screen instructions.