Make sure the following requirements are fulfilled:
  • The built-in Windows Firewall service must be running. If you need to turn off the Windows Firewall for some reason, use the Windows Firewall Control Panel program (Control Panel > System and Security > Windows Firewall), but leave the Windows Firewall service running.
  • Ensure that the person performing the installation has local administrator privileges in Windows. Reboot the computer if you are asked to do so. After the reboot, the administrator who started the setup must log in again so that the installation process can be completed correctly.
  • Ensure that third-party firewalls are turned off.
CAUTION
Make sure that there is enough free drive space on your PC. Otherwise, the installation will fail.
Setup time varies depending on the presence of third-party components. For example, installing .NET Framework libraries may take a considerable amount of time.