If your Chromeleon installation (versions older than Chromeleon 7.2 SR1) includes software development kit (SDK) or driver development kit (DDK) add-ins that are installed using a third-party setup, the existing SDK and DDK add-ins will not be moved to the new Thermo installation location.
To make the DDK and SDK add-ins available in Chromeleon 7.2 SR1 and later:
Procedure
  1. Uninstall the DDK and SDK drivers before you upgrade to Chromeleon 7.2 SR1 and later.
  2. Upgrade Chromeleon.
  3. Reinstall the DDK and SDK drivers again.
Alternatively, you can change the Chromeleon installation folder to Program Files (x86)\Dionex\Chromeleon. For more information, see Install Chromeleon 7 – main and service release.